Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011680.2 | NP_035810.1 | 3 - 4 | 274 | 93 | |
GenBank mRNA | AB221562.1 | - | 3 - 4 | 233 | 93 | |
AK146871.1 | - | 3 - 4 | 221 | 93 | ||
BC082995.1 | - | 3 - 4 | 274 | 6440967 | 93 | |
U01662.1 | - | 3 - 4 | 290 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
