Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900005808.mm.1 |
TC0900000776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005808.mm.1 |
NM_001083927.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005808.mm.1 |
NM_001083927.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005808.mm.1 |
NM_001083928.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005808.mm.1 |
NM_001083928.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005808.mm.1 |
NM_009389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005808.mm.1 |
NM_009389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005813.mm.1 |
TC0900000776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005813.mm.1 |
NM_001083927.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005813.mm.1 |
NM_001083927.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005813.mm.1 |
NM_001083928.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005813.mm.1 |
NM_001083928.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005813.mm.1 |
NM_009389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005813.mm.1 |
NM_009389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005809.mm.1 |
TC0900000776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005809.mm.1 |
NM_001083927.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005809.mm.1 |
NM_001083927.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005809.mm.1 |
NM_001083928.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005809.mm.1 |
NM_001083928.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005809.mm.1 |
NM_009389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005809.mm.1 |
NM_009389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005814.mm.1 |
TC0900000776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005814.mm.1 |
NM_001083927.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005814.mm.1 |
NM_001083927.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005814.mm.1 |
NM_001083928.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005814.mm.1 |
NM_001083928.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005814.mm.1 |
NM_009389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005814.mm.1 |
NM_009389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005807.mm.1 |
TC0900000776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005807.mm.1 |
NM_001083927.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005807.mm.1 |
NM_001083927.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005807.mm.1 |
NM_001083928.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005807.mm.1 |
NM_001083928.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005807.mm.1 |
NM_009389.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005807.mm.1 |
NM_009389.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005810.mm.1 |
TC0900000776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005810.mm.1 |
NM_001083927.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005810.mm.1 |
NM_001083927.1 |
Refseq |