Assay Details
Gene Symbol: Cyp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009993.3 | NP_034123.1 | 1 - 2 | 47 | 63 | |
GenBank mRNA | BC054827.1 | - | 1 - 2 | 40 | 5054733 | 63 |
K02589.1 | - | 1 - 2 | 48 | 63 | ||
X00479.1 | - | 1 - 2 | 48 | 63 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC054827.1, K02589.1, NM_009993.3, X00479.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
