Assay Details
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153406.3 | NP_700455.3 | 1 - 2 | 142 | 63 | |
GenBank mRNA | AK035500.1 | - | 1 - 2 | 64 | ||
AK052716.1 | - | 1 - 2 | 143 | 63 | ||
AK172502.1 | - | 1 - 2 | 120 | 63 | ||
AY884297.1 | - | 1 - 2 | 143 | 63 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK052716.1, AY884297.1, NM_153406.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
