Assay Details
Gene Symbol: Tmprss9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081688.2 | NP_001075157.2 | 17 - 18 | 3141 | 109 | |
GenBank mRNA | BC158102.1 | - | 17 - 18 | 3237 | 9007387 | 109 |
BC172169.1 | - | 17 - 18 | 3237 | 9054839 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976949 | AB109391.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976953 | NM_001081688.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906217 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16121 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500394 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629095 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530696 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788835 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307547 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608342 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726643 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124435 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100687 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976953 | NM_001081688.2 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27286070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08476904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08354259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08254734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08628876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08707481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08670391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08670394 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08767622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08690046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08936394 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286194 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08527856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08887318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08672262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08767629 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08433696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08773651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08392838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08672269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08339748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08707478 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08392836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08773653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08783259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08340631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08670392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08887315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08660698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286187 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08276998 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08767623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08672264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27286065 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10364935 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364943 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364934 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364946 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364941 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364944 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364936 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364947 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364949 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364948 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364933 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364945 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364940 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364942 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10364932 | - | - |
Affymetrix Mouse Clariom S Array | PSR1000006759.mm.1 | TC1000000875.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006759.mm.1 | NM_001081688.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006759.mm.1 | NM_001081688.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006758.mm.1 | TC1000000875.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006758.mm.1 | NM_001081688.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006758.mm.1 | NM_001081688.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006760.mm.1 | TC1000000875.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006760.mm.1 | NM_001081688.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006760.mm.1 | NM_001081688.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 361317 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_3142104 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3142104 | - | - |
