Assay Details
Gene Symbol: Cdon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021339.2 | NP_067314.2 | 11 - 12 | 2376 | 102 | |
GenBank mRNA | AF090866.1 | - | 11 - 12 | 2376 | 102 | |
BC109176.2 | - | 11 - 12 | 2375 | 40048829 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
