Assay Details
Gene Symbol: Zmym6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285887.1 | NP_001272816.1 | 1 - 2 | 225 | 68 | |
NM_001285888.1 | NP_001272817.1 | 1 - 2 | 225 | 68 | ||
NM_177462.5 | NP_803413.1 | 1 - 2 | 225 | 68 | ||
GenBank mRNA | AK035136.1 | - | 1 - 2 | 227 | 68 | |
BC140958.1 | - | 1 - 2 | 100 | 9055570 | 68 | |
BC144807.1 | - | 1 - 2 | 100 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK035136.1, BC140958.1, NM_001285887.1, NM_001285888.1, NM_177462.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
