Assay Details
Gene Symbol: Zmym6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285885.1 | NP_001272814.1 | 2 - 3 | 1232 | 143 | |
NM_001285887.1 | NP_001272816.1 | 3 - 4 | 425 | 143 | ||
NM_177462.5 | NP_803413.1 | 3 - 4 | 425 | 143 | ||
NR_104366.1 | 2 - 3 | 1232 | 143 | |||
GenBank mRNA | AK035136.1 | - | 3 - 4 | 427 | 143 | |
AK081836.1 | - | 3 - 4 | 415 | 143 | ||
BC094598.1 | - | 2 - 3 | 1205 | 143 | ||
BC140958.1 | - | 3 - 4 | 300 | 9055570 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
