Assay Details
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172495.5 | NP_766083.3 | 2 - 3 | 397 | 74 | |
GenBank mRNA | AK128935.1 | - | 1 - 2 | 219 | 75 | |
AK144324.1 | - | 2 - 3 | 229 | 74 | ||
AK166129.1 | - | 2 - 3 | 619 | 74 | ||
BB870337.1 | - | 2 - 3 | 229 | 74 | ||
BC141150.1 | - | 1 - 2 | 131 | 9055762 | 74 | |
BC145459.1 | - | 1 - 2 | 131 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK128935.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
