Assay Details
Gene Symbol: Kdm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133872.2 | NP_598633.2 | 15 - 16 | 2127 | 69 | |
GenBank mRNA | AK007763.1 | - | 2 - 3 | 335 | 69 | |
AK129170.1 | - | 15 - 16 | 2067 | 69 | ||
BC019417.1 | - | 1 - 2 | 73 | 69 | ||
BC059885.1 | - | 15 - 16 | 2114 | 6815975 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
