Assay Details
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007595.5 | NP_031621.3 | 14 - 15 | 1275 | 85 | |
GenBank mRNA | AK046454.1 | - | 14 - 15 | 1276 | 85 | |
AK163698.1 | - | 14 - 15 | 1092 | 85 | ||
X63615.1 | - | 14 - 15 | 1090 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
