Assay Details
Gene Symbol: Nr2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011630.3 | NP_035760.1 | 5 - 6 | 787 | 62 | |
GenBank mRNA | AK157305.1 | - | 5 - 6 | 761 | 62 | |
BC103683.1 | - | 3 - 4 | 406 | 40047943 | 62 | |
BC103684.1 | - | 3 - 4 | 406 | 40047942 | 62 | |
BC103686.1 | - | 3 - 4 | 406 | 40047941 | 62 | |
S75970.1 | - | 5 - 6 | 902 | 62 | ||
U11688.1 | - | 4 - 5 | 449 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
