Assay Details
Gene Symbol: Mcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008564.2 | NP_032590.2 | 2 - 3 | 369 | 77 | |
GenBank mRNA | AK037751.1 | - | 2 - 3 | 293 | 77 | |
AK077801.1 | - | 2 - 3 | 271 | 77 | ||
AK088156.1 | - | 2 - 3 | 282 | 77 | ||
AK088378.1 | - | 2 - 3 | 269 | 77 | ||
AK129039.1 | - | 2 - 3 | 250 | 77 | ||
AK146187.1 | - | 2 - 3 | 284 | 77 | ||
AK146375.1 | - | 2 - 3 | 293 | 77 | ||
BC055318.1 | - | 2 - 3 | 278 | 5696348 | 77 | |
D86725.1 | - | 2 - 3 | 289 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
