Assay Details
Gene Symbol: Usp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013700.1 | NP_038728.1 | 16 - 17 | 2154 | 77 | |
GenBank mRNA | AK154008.1 | - | 16 - 17 | 2074 | 77 | |
BC066993.1 | - | 16 - 17 | 2132 | 5689229 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
