Assay Details
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033126.2 | NP_001028298.1 | 1 - 2 | 266 | 104 | |
NM_001286753.1 | NP_001273682.1 | 1 - 2 | 266 | 104 | ||
GenBank mRNA | AA475779.1 | - | 1 - 2 | 193 | 104 | |
AK137770.1 | - | 1 - 2 | 243 | 104 | ||
AK153895.1 | - | 1 - 2 | 205 | 104 | ||
AK154005.1 | - | 1 - 2 | 250 | 104 | ||
BC139125.1 | - | 1 - 2 | 231 | 8862147 | 104 | |
BC139126.1 | - | 1 - 2 | 231 | 8862148 | 104 | |
DT917063.1 | - | 1 - 2 | 192 | 104 | ||
DT924276.1 | - | 1 - 2 | 192 | 104 | ||
L24495.1 | - | 1 - 2 | 308 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
