Assay Details
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033126.2 | NP_001028298.1 | 2 - 3 | 398 | 89 | |
GenBank mRNA | AA475779.1 | - | 2 - 3 | 324 | 88 | |
AK153895.1 | - | 2 - 3 | 337 | 89 | ||
AK154005.1 | - | 2 - 3 | 382 | 89 | ||
BC139125.1 | - | 2 - 3 | 363 | 8862147 | 89 | |
BC139126.1 | - | 2 - 3 | 363 | 8862148 | 89 | |
DT917063.1 | - | 2 - 3 | 324 | 89 | ||
L24495.1 | - | 2 - 3 | 440 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
