Assay Details
Gene Symbol: Egfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007912.4 | NP_031938.1 | 10 - 11 | 1488 | 110 | |
NM_207655.2 | NP_997538.1 | 10 - 11 | 1488 | 110 | ||
GenBank mRNA | AF124513.1 | - | 10 - 11 | 1422 | 110 | |
AF275367.1 | - | 10 - 11 | 1431 | 110 | ||
AK004883.1 | - | 10 - 11 | 1489 | 110 | ||
AK004911.1 | - | 10 - 11 | 1463 | 110 | ||
AK004944.1 | - | 10 - 11 | 1482 | 110 | ||
AK134262.1 | - | 10 - 11 | 1461 | 110 | ||
BC023729.1 | - | 10 - 11 | 1459 | 5351807 | 110 | |
U03425.1 | - | 10 - 11 | 1258 | 110 | ||
X59698.1 | - | 10 - 11 | 1367 | 110 | ||
X78987.1 | - | 10 - 11 | 1267 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
