Assay Details
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025958.2 | NP_080234.2 | 10 - 11 | 2992 | 61 | |
GenBank mRNA | AK129186.1 | - | 10 - 11 | 2963 | 61 | |
AK145112.1 | - | 10 - 11 | 2977 | 61 | ||
AK170420.1 | - | 1 - 2 | 696 | 61 | ||
BC056365.1 | - | 10 - 11 | 2974 | 6824847 | 61 | |
BC058338.1 | - | 9 - 10 | 2807 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
