Assay Details
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025958.2 | NP_080234.2 | 11 - 12 | 3093 | 59 | |
GenBank mRNA | AK129186.1 | - | 11 - 12 | 3064 | 59 | |
AK145112.1 | - | 11 - 12 | 3078 | 59 | ||
AK170420.1 | - | 2 - 3 | 797 | 59 | ||
BC056365.1 | - | 11 - 12 | 3075 | 6824847 | 59 | |
BC058338.1 | - | 10 - 11 | 2908 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
