Assay Details
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025958.2 | NP_080234.2 | 2 - 3 | 268 | 76 | |
GenBank mRNA | AK129186.1 | - | 2 - 3 | 239 | 76 | |
AK145112.1 | - | 2 - 3 | 253 | 76 | ||
BC056365.1 | - | 2 - 3 | 250 | 6824847 | 76 | |
BC058338.1 | - | 1 - 2 | 83 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
