Assay Details
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025958.2 | NP_080234.2 | 4 - 5 | 543 | 143 | |
GenBank mRNA | AK129186.1 | - | 4 - 5 | 514 | 143 | |
AK145112.1 | - | 4 - 5 | 528 | 143 | ||
AK214895.1 | - | 1 - 2 | 130 | 143 | ||
BC056365.1 | - | 4 - 5 | 525 | 6824847 | 143 | |
BC058338.1 | - | 3 - 4 | 358 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
