Assay Details
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025958.2 | NP_080234.2 | 9 - 10 | 1490 | 63 | |
GenBank mRNA | AK129186.1 | - | 9 - 10 | 1461 | 63 | |
AK145112.1 | - | 9 - 10 | 1475 | 63 | ||
BC020469.1 | - | 2 - 3 | 230 | 63 | ||
BC056365.1 | - | 9 - 10 | 1472 | 6824847 | 63 | |
BC058338.1 | - | 8 - 9 | 1305 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
