Assay Details
Gene Symbol: Fig4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133999.1 | NP_598760.1 | 4 - 5 | 628 | 142 | |
GenBank mRNA | AK041196.1 | - | 4 - 5 | 629 | 142 | |
AK155392.1 | - | 4 - 5 | 625 | 142 | ||
AK169881.1 | - | 4 - 5 | 592 | 142 | ||
AK172130.1 | - | 4 - 5 | 622 | 142 | ||
AK172341.1 | - | 4 - 5 | 592 | 143 | ||
BC015295.1 | - | 4 - 5 | 628 | 4221630 | 142 | |
BC031887.1 | - | 4 - 5 | 627 | 5363304 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
