Assay Details
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033126.2 | NP_001028298.1 | 4 - 5 | 643 | 82 | |
NM_001042564.1 | NP_001036029.1 | 2 - 3 | 331 | 82 | ||
NM_001286753.1 | NP_001273682.1 | 3 - 4 | 463 | 82 | ||
GenBank mRNA | AI326735.1 | - | 2 - 3 | 331 | 82 | |
AK153895.1 | - | 4 - 5 | 582 | 82 | ||
AK154005.1 | - | 4 - 5 | 627 | 82 | ||
BC139125.1 | - | 4 - 5 | 608 | 8862147 | 82 | |
BC139126.1 | - | 4 - 5 | 608 | 8862148 | 82 | |
BC171931.1 | - | 2 - 3 | 296 | 9054107 | 82 | |
BE631582.1 | - | 3 - 4 | 282 | 82 | ||
DT924276.1 | - | 3 - 4 | 389 | 82 | ||
L24495.1 | - | 4 - 5 | 685 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
