Assay Details
Gene Symbol: Gm11999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040327.1 | - | 2 - 3 | 187 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930512M02 gene | 4930512M02Rik | -25.4kb | Sense | Chr. 11 - 11589223 - 11627818 on NCBI Build GRCm38 | |
Downstream | IKAROS family zinc finger 1 | Ikzf1 |
5832432G11Rik, Ikaros, LyF-1, Zfpn1a1, Znfn1a1, hlk-1, mKIAA4227 |
31.3kb | Anti-sense | Chr. 11 - 11684969 - 11772926 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
