Assay Details
Gene Symbol: Myct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026793.2 | NP_081069.1 | 1 - 2 | 171 | 60 | |
GenBank mRNA | AK030706.1 | - | 1 - 2 | 96 | 60 | |
AK054222.1 | - | 1 - 2 | 70 | 60 | ||
AK142963.1 | - | 1 - 2 | 104 | 60 | ||
AY090114.1 | - | 1 - 2 | 125 | 60 | ||
BB624713.1 | - | 1 - 2 | 171 | 60 | ||
BC096559.1 | - | 1 - 2 | 121 | 1399480 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
