Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 10 - 11 | 2641 | 68 | |
NM_153787.2 | NP_722482.1 | 10 - 11 | 2635 | 68 | ||
GenBank mRNA | AK079110.1 | - | 6 - 7 | 891 | 68 | |
AK129071.1 | - | 7 - 8 | 1925 | 68 | ||
AK149724.1 | - | 10 - 11 | 2594 | 68 | ||
AK150017.1 | - | 10 - 11 | 2594 | 68 | ||
BC029682.1 | - | 10 - 11 | 2573 | 68 | ||
BC032973.1 | - | 1 - 2 | 316 | 68 | ||
BC034300.1 | - | 10 - 11 | 2567 | 4507876 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
