Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 1 - 2 | 134 | 129 | |
NM_001025393.1 | NP_001020564.1 | 1 - 2 | 134 | 129 | ||
NM_153787.2 | NP_722482.1 | 1 - 2 | 134 | 129 | ||
GenBank mRNA | AK011802.1 | - | 1 - 2 | 74 | 129 | |
AK088741.1 | - | 1 - 2 | 75 | 129 | ||
AK141834.1 | - | 1 - 2 | 87 | 129 | ||
AK149724.1 | - | 1 - 2 | 87 | 129 | ||
AK150017.1 | - | 1 - 2 | 87 | 129 | ||
AK161692.1 | - | 1 - 2 | 132 | 129 | ||
AK162896.1 | - | 1 - 2 | 92 | 129 | ||
BC028963.1 | - | 1 - 2 | 66 | 129 | ||
BC029682.1 | - | 1 - 2 | 65 | 129 | ||
BC034300.1 | - | 1 - 2 | 66 | 4507876 | 129 | |
BC086624.1 | - | 1 - 2 | 119 | 129 | ||
BY071148.1 | - | 1 - 2 | 135 | 129 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK011802.1, AK088741.1, AK141834.1, AK149724.1, AK150017.1, AK161692.1, AK162896.1, BC028963.1, BC034300.1, NM_001025392.1, NM_001025393.1, NM_153787.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
