Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 2 - 3 | 143 | ||
NM_001025393.1 | NP_001020564.1 | 2 - 3 | 143 | |||
NM_153787.2 | NP_722482.1 | 2 - 3 | 143 | |||
GenBank mRNA | AK011802.1 | - | 2 - 3 | 143 | ||
AK088741.1 | - | 2 - 3 | 143 | |||
AK141834.1 | - | 2 - 3 | 143 | |||
AK149724.1 | - | 2 - 3 | 143 | |||
AK150017.1 | - | 2 - 3 | 143 | |||
AK161692.1 | - | 2 - 3 | 143 | |||
AK162896.1 | - | 2 - 3 | 143 | |||
BC028963.1 | - | 2 - 3 | 143 | |||
BC029682.1 | - | 2 - 3 | 143 | |||
BC034300.1 | - | 2 - 3 | 4507876 | 143 | ||
BC086624.1 | - | 2 - 3 | 143 | |||
BY071148.1 | - | 2 - 3 | 143 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK011802.1, AK088741.1, AK141834.1, AK149724.1, AK150017.1, AK161692.1, AK162896.1, BC028963.1, BC034300.1, NM_001025392.1, NM_001025393.1, NM_153787.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
