Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 5 - 6 | 1927 | 65 | |
NM_001025393.1 | NP_001020564.1 | 5 - 6 | 1927 | 65 | ||
NM_153787.2 | NP_722482.1 | 5 - 6 | 1921 | 65 | ||
GenBank mRNA | AK079110.1 | - | 1 - 2 | 177 | 65 | |
AK088741.1 | - | 5 - 6 | 1862 | 65 | ||
AK129071.1 | - | 2 - 3 | 1211 | 65 | ||
AK141834.1 | - | 5 - 6 | 1874 | 65 | ||
AK149724.1 | - | 5 - 6 | 1880 | 65 | ||
AK150017.1 | - | 5 - 6 | 1880 | 65 | ||
AK161608.1 | - | 817 | 65 | |||
AK161692.1 | - | 5 - 6 | 1925 | 65 | ||
BC029682.1 | - | 5 - 6 | 1858 | 65 | ||
BC034300.1 | - | 5 - 6 | 1853 | 4507876 | 65 | |
BC086624.1 | - | 5 - 6 | 1909 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
