Assay Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133681.4 | NP_598442.1 | 1 - 2 | 135 | 111 | |
GenBank mRNA | AK007428.1 | - | 1 - 2 | 135 | 111 | |
AK078869.1 | - | 1 - 2 | 136 | 111 | ||
AK162521.1 | - | 1 - 2 | 136 | 111 | ||
BC003448.1 | - | 1 - 2 | 75 | 2654254 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK078869.1, BC003448.1, NM_133681.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
