Assay Details
Gene Symbol: Ddc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016672.4 | NP_057881.1 | 1 - 2 | 71 | 82 | |
GenBank mRNA | AK149141.1 | - | 1 - 2 | 65 | 82 | |
AK160769.1 | - | 1 - 2 | 65 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK149141.1, AK160769.1, NM_016672.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
