Assay Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008917.3 | NP_032943.2 | 2 - 3 | 263 | 92 | |
GenBank mRNA | AF071025.1 | - | 2 - 3 | 280 | 92 | |
AF087568.1 | - | 2 - 3 | 241 | 92 | ||
AF326557.1 | - | 2 - 3 | 247 | 92 | ||
AF326558.1 | - | 2 - 3 | 247 | 92 | ||
AK014561.1 | - | 2 - 3 | 245 | 92 | ||
AK148314.1 | - | 2 - 3 | 238 | 92 | ||
AK151055.1 | - | 2 - 3 | 238 | 92 | ||
AK151107.1 | - | 2 - 3 | 249 | 92 | ||
AK152474.1 | - | 2 - 3 | 247 | 92 | ||
AK152940.1 | - | 2 - 3 | 249 | 92 | ||
AK153106.1 | - | 2 - 3 | 244 | 92 | ||
AK153316.1 | - | 2 - 3 | 244 | 92 | ||
AK169579.1 | - | 3 - 4 | 338 | 92 | ||
AK169658.1 | - | 3 - 4 | 344 | 92 | ||
AK171668.1 | - | 2 - 3 | 245 | 92 | ||
CB600397.1 | - | 2 - 3 | 266 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
