Assay Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008917.3 | NP_032943.2 | 4 - 5 | 461 | 87 | |
GenBank mRNA | AF071025.1 | - | 4 - 5 | 478 | 87 | |
AF087568.1 | - | 4 - 5 | 439 | 87 | ||
AF326557.1 | - | 4 - 5 | 445 | 87 | ||
AF326558.1 | - | 4 - 5 | 445 | 87 | ||
AK014561.1 | - | 4 - 5 | 443 | 87 | ||
AK148314.1 | - | 4 - 5 | 436 | 87 | ||
AK151055.1 | - | 4 - 5 | 436 | 87 | ||
AK151107.1 | - | 4 - 5 | 447 | 87 | ||
AK152474.1 | - | 4 - 5 | 445 | 87 | ||
AK152940.1 | - | 4 - 5 | 447 | 87 | ||
AK153106.1 | - | 4 - 5 | 442 | 87 | ||
AK153316.1 | - | 4 - 5 | 442 | 87 | ||
AK169579.1 | - | 5 - 6 | 536 | 87 | ||
AK169658.1 | - | 5 - 6 | 542 | 87 | ||
AK171668.1 | - | 4 - 5 | 443 | 87 | ||
AK192077.1 | - | 2 - 3 | 116 | 87 | ||
AK192572.1 | - | 2 - 3 | 116 | 87 | ||
AK216167.1 | - | 2 - 3 | 113 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
