Assay Details
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011282.2 | NP_035412.2 | 20 - 21 | 3252 | 96 | |
GenBank mRNA | U15443.1 | - | 20 - 21 | 3251 | 96 | |
X81650.1 | - | 20 - 21 | 3075 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
