Assay Details
Gene Symbol: Il20ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172786.2 | NP_766374.1 | 3 - 4 | 463 | 77 | |
GenBank mRNA | AK054215.1 | - | 3 - 4 | 464 | 77 | |
BC056628.1 | - | 3 - 4 | 447 | 5033177 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
