Assay Details
Gene Symbol: Tbc1d32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033385.3 | NP_001028557.2 | 18 - 19 | 2204 | 75 | |
GenBank mRNA | AK141206.1 | - | 3 - 4 | 399 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
