Assay Details
Gene Symbol: Reps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111065.1 | NP_001104535.1 | 12 - 13 | 1541 | 66 | |
NM_009048.2 | NP_033074.2 | 13 - 14 | 1622 | 66 | ||
GenBank mRNA | AF031939.1 | - | 12 - 13 | 1443 | 66 | |
AK150284.1 | - | 13 - 14 | 1486 | 66 | ||
AK151309.1 | - | 13 - 14 | 1486 | 66 | ||
BC002256.1 | - | 2 - 3 | 119 | 66 | ||
BC087547.1 | - | 12 - 13 | 1489 | 30441027 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
