Assay Details
Gene Symbol: Reps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111065.1 | NP_001104535.1 | 1 - 2 | 175 | 152 | |
NM_009048.2 | NP_033074.2 | 1 - 2 | 175 | 152 | ||
GenBank mRNA | BC087547.1 | - | 1 - 2 | 123 | 30441027 | 152 |
BF318674.1 | - | 1 - 2 | 175 | 152 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC087547.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
