Assay Details
Gene Symbol: Akap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK085630.1 | - | 1 - 2 | 149 | 100 | |
AK139238.1 | - | 1 - 2 | 166 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
