Assay Details
Gene Symbol: Artn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284191.1 | NP_001271120.1 | 3 - 4 | 652 | 61 | |
GenBank mRNA | AK015393.1 | - | 3 - 4 | 360 | 61 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK015393.1, NM_001284191.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
