Assay Details
Gene Symbol: A1cf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081074.1 | NP_001074543.1 | 7 - 8 | 1013 | 79 | |
GenBank mRNA | AK007970.1 | - | 6 - 7 | 857 | 79 | |
AY566862.1 | - | 7 - 8 | 1010 | 79 | ||
AY566863.1 | - | 7 - 8 | 1010 | 79 | ||
AY566864.1 | - | 7 - 8 | 1010 | 79 | ||
BC038942.1 | - | 5 - 6 | 713 | 79 | ||
BC089622.1 | - | 7 - 8 | 961 | 6760455 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
