Assay Details
Gene Symbol: Taf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081288.1 | NP_001074757.1 | 10 - 11 | 1504 | 85 | |
GenBank mRNA | AK028828.1 | - | 10 - 11 | 1251 | 85 | |
AK147903.1 | - | 10 - 11 | 1506 | 85 | ||
BC147638.1 | - | 10 - 11 | 1455 | 9056628 | 85 | |
BC151079.1 | - | 10 - 11 | 1455 | 9087992 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
