Assay Details
Gene Symbol: Taf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081288.1 | NP_001074757.1 | 20 - 21 | 2923 | 88 | |
GenBank mRNA | AK147903.1 | - | 20 - 21 | 2926 | 88 | |
BC147638.1 | - | 20 - 21 | 2874 | 9056628 | 88 | |
BC151079.1 | - | 20 - 21 | 2874 | 9087992 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
