Assay Details
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF229633.1 | - | 12 - 13 | 1078 | 126 | |
AK122481.2 | - | 10 - 11 | 937 | 126 | ||
AK134989.1 | - | 12 - 13 | 1712 | 126 | ||
BC086631.1 | - | 13 - 14 | 1622 | 126 | ||
BU149633.1 | - | 116 | 126 | |||
U61363.1 | - | 4 - 5 | 481 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
