Assay Details
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168250.1 | NP_001161722.1 | 5 - 6 | 693 | 116 | |
NM_134092.3 | NP_598853.3 | 5 - 6 | 693 | 116 | ||
GenBank mRNA | AK080036.1 | - | 5 - 6 | 507 | 116 | |
AK088549.1 | - | 5 - 6 | 507 | 116 | ||
AK133855.1 | - | 5 - 6 | 541 | 116 | ||
BC052197.1 | - | 5 - 6 | 693 | 6511752 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
