Assay Details
Gene Symbol: Ap4m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021392.4 | NP_067367.3 | 8 - 9 | 842 | 89 | |
GenBank mRNA | AF242858.1 | - | 8 - 9 | 710 | 89 | |
AK031638.1 | - | 8 - 9 | 845 | 89 | ||
BC011174.1 | - | 8 - 9 | 727 | 4207033 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
