Assay Details
Gene Symbol: Taok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081308.2 | NP_001074777.1 | 17 - 18 | 2418 | 80 | |
NM_001199685.1 | NP_001186614.1 | 18 - 19 | 2526 | 80 | ||
NM_183306.2 | NP_899129.2 | 18 - 19 | 2503 | 80 | ||
GenBank mRNA | AK039665.1 | - | 5 - 6 | 1076 | 80 | |
AK217809.1 | - | 1 - 2 | 37 | 80 | ||
BC158072.1 | - | 18 - 19 | 2468 | 9088167 | 80 | |
CK625948.1 | - | 3 - 4 | 475 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
