Assay Details
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141813.1 | - | 11 - 12 | 1522 | 66 | |
AK170166.1 | - | 10 - 11 | 1292 | 66 | ||
AK172083.1 | - | 10 - 11 | 1281 | 66 | ||
AK207938.1 | - | 2 - 3 | 79 | 66 | ||
AK218253.1 | - | 2 - 3 | 79 | 66 | ||
BC063768.1 | - | 11 - 12 | 1502 | 66 | ||
U14166.1 | - | 11 - 12 | 1545 | 66 | ||
U15159.1 | - | 11 - 12 | 1517 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
