Assay Details
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141813.1 | - | 14 - 15 | 1807 | 106 | |
AK170166.1 | - | 14 - 15 | 2133 | 106 | ||
AK172083.1 | - | 13 - 14 | 1566 | 106 | ||
BC063768.1 | - | 14 - 15 | 1787 | 106 | ||
BQ828670.1 | - | 2 - 3 | 174 | 106 | ||
U14166.1 | - | 14 - 15 | 1830 | 106 | ||
U15159.1 | - | 14 - 15 | 1802 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
